VWR® Formaldehyde-Free Sheep Brains
Supplier: VWR International
Certificates
About this item
For a 100% formaldehyde-free dissection experience, these form-free preserved specimens have never been touched by any formaldehyde whatsoever.
- 100% form-free
- Safe for students
For those whose main priority is avoiding formaldehyde completely, Ward’s offers this alternative to traditionally preserved specimens. These specimens have never been exposed to formaldehyde. They are preserved using a proprietary formulation which binds with the tissue as non-toxic, cross-linked proteins. However, for true-to-life, high quality preservation that maintains the best detail, we recommend upgrading to our Ward’s Pure preserved specimens instead.
Frequently Bought Together