Urinary Indican Test Kit, BioAssay Systems
Product Details & Documents
The Urinary Indican Test Kit is an accurate, simple, fast (15 minutes), and inexpensive screening tool for the identification of protein putrefaction in the gastrointestinal tract.
- Convenient design: simply pipette 2 ml urine into the ready reagent vial, mix, and read the indican level from a color chart
- Fast results available in 15 minutes
- Method: Obermeyer
Indican (indoxyl sulfate) is a putrefaction product resulting from bacterial deconjugation of dietary tryptophan to indole in the small intestine. Indican levels are directly associated with bacterial activity in the intestines. Elevated levels indicate intestinal toxemia or overgrowth of anaerobic bacteria, putrefaction of undigested food in the bowels, stomach disorder (constipation, malabsorption), intestinal disorders, and pancreatic insufficiency.
BioAssay Systems' Urinary Indican Test Kit is a less hazardous modification of the original Obermeyer indican reagent. It provides an accurate, simple, fast, and inexpensive screening tool for the identification of protein putrefaction in the gastrointestinal tract.
: For research use only. Not intended for diagnostic use. The indican reagent is corrosive.
Specifications
Specifications
- Description:Urinary Indican Test Kit
- No. of tests:20