


Papanicolaou's solution EA 36, VWR® stain for histology, for cytology
Catalog # 10143-156
Supplier: VWR International
CAS Number:
Specifications
- Size:3.8 L (1 gal.)
- Cat. No.:10143-156
Specifications
Specification Test Results
- Absorbance (Visible Scan 700- 400nm) @626 nm:370 – 500 MA
- Absorbance (Visible Scan 700- 400nm) @ 528nm:270 – 350 MA
Specification Test Results
About this item
:
UN:1170
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient