Order Entry
United States
ContactUsLinkComponent
 
PM Perfection Receipt Paper, White, Eaton Office Supply
Catalog # 470150-092
PM Perfection Receipt Paper, White, Eaton Office Supply
Catalog # 470150-092
Supplier Number:  PMC 05208

Specifications

  • For Use With:
    Omniprint OM20, Seiko, DPU201G, DPU20, DPU212, SPT211-144, STP211192 and LTP251
  • Color:
    White
  • Description:
    Machine receipt thermal print rolls, 50 per carton
  • Dimensions:
    2.25×80"
  • Cat. No.:
    470150-092
  • Supplier no.:
    PMC 05208

Specifications

About this item

Quality thermal paper for use with cash register/ATM rolls.

  • Machine receipt print rolls
  • Cash register paper
  • ATM paper
  • Works efficiently with your machine printhead

Cash Register/ATM Rolls offer top-quality thermal paper that works efficiently with your machine printhead. High quality rolls provide crisp, clear black images. Designed for use in Omniprint OM20, Seiko, DPU201G, DPU20, DPU212, SPT211-144, STP211-192 and LTP251.