


Titration solvent ASTM D-2896, LabReady™, Solvent Blend total base number titration solvent, Burdick & Jackson™
Catalog # BJWP2151-19P
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Winpak®
- Size:19 L
- Cat. No.:BJWP2151-19P
Specifications
About this item
:
Boiling Pt:115,6 °C
Density:1,0795 g/cm3 (20 °C)
Flash Pt:7,2 °C
Storage Temperature:Ambient
UN:2924
ADR:3,III
:GHS07
:GHS05
:GHS02