

tert-Butyl methyl ether ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Catalog # BJLP242-4
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJLP242-4
Specifications
Specification Test Results
- Appearance:Passes test
- Assay by GC Analysis:99 % min.
- Color (APHA):10 max.
- Peroxides (as H₂O₂) at time of packaging:1 ppm max.
- Residue after Evaporation:0.001 % max.
- Water by Karl Fischer Titration:0.05 % max.
- SolubilityinWater(%)at20°C:4.8
- SurfaceTension(dyn/cm)at20°C:19.4
- VaporPressure(Torr)at20°C:240
Specification Test Results
About this item
:
:88.15 g/mol
Boiling Pt:15.1 %(V)
Melting Pt:–108.6 °C
Density:0.74 g/cm³ (20 °C)
Flash Pt:–27 °C
UN:2398
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812