Order Entry
United States
ContactUsLinkComponent
 
Irritant
Health
Flammable
N,N-Dimethylformamide ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Catalog # BJLP076-19
CAS Number:  
undefined
Irritant
Health
Flammable
N,N-Dimethylformamide ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Catalog # BJLP076-19
Supplier Number:  LP076-19
CAS Number:  

Specifications

  • Pack type:
    Metal can
  • Size:
    19 L
  • Cat. No.:
    BJLP076-19

Specifications

Specification Test Results

  • Appearance:
    Passes test
  • Assay by GC Analysis:
    99.8 % min.
  • Color (APHA):
    10 max.
  • Residue after Evaporation:
    0.005 % max.
  • Titrable Acid:
    0.5 µeq/g max.
  • Titrable Base:
    3 µeq/g max.
  • Water by Karl Fischer Titration:
    0.15 % max.
  • VaporPressure(Torr)at20°C:
    2.7

Specification Test Results

About this item

 :
 :73.09 g/mol
Boiling Pt:15.2 %(V)
Melting Pt:–60.4 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:58 °C
UN:2265
ADR:3,III
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284