


p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
BJ95682-2.5L
:
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:BJ95682-2.5L
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.494 - 1.496
- Non-volatile matter:Max. 0.005 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- o-Xylene (GC):Max. 0.5 %
- m-Xylole (GC):Max. 0.5 %
- Sum of the isomers (GC):Max. 0.5 %
- Benzene (GC):Max. 1.0 %
- Ethyl benzene (GC):Max. 1.0 %
- Toluene (GC):Max. 0.5 %
- Refractive Index:n20/D 1.495
:
:106.17 g/mol
:137-139 °C (1.013 hPa)
:137…139 °C (1.013 hPa)
:12 °C
:0.86 g/cm³ (20 °C)
:25 °C
:Ambient
:203-396-5
:1307
:3,III
:GHS02
:GHS08
:GHS07
:106-42-3
:C₆H₄(CH₃)₂
:MFCD00008556