Order Entry
United States
ContactUsLinkComponent
Acetonitrile : Water (25:75)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile : Water (25:75)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ803-4
Supplier:  Avantor
CAS Number:  
Acetonitrile : Water (25:75)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ803-4
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    BJ803-4

Specifications

About this item

 :
Boiling Pt:70 °C
Density:0.9594 g/cm³ (25 °C)
Flash Pt:13.9 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
CAS Number:75-05-8
 :7732-18-5