Order Entry
United States
ContactUsLinkComponent
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
Catalog # BJ679178-4X4L
CAS Number:  
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
Catalog # BJ679178-4X4L
Supplier Number:  679178-4X4L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4x 4 L
  • Cat. No.:
    BJ679178-4X4L

Specifications

Specification Test Results

  • Contains:
    0.05 %(w/v) ammonium formate

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Density:0.9986 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
 :64-18-6
MDL Number:MFCD00011332