
Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™
Catalog # BJ656054-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ656054-1L
Specifications
Specification Test Results
- Assay:9.80 - 10.20 N
- Carbonate (as Na₂CO₃):Max. 0.12 N
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:120 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548