Water with 0,1% (v/v) trifluoroacetic acid 0.05% in aqueous solution, CHROMASOLV™, Solvent Blend for HPLC, Riedel-de Haën™
Catalog # BJ590142-4X4L
Supplier: Honeywell Research Chemicals
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:4x 4 L
- Cat. No.:BJ590142-4X4L
Specifications
Specification Test Results
- Contains:0.05 %(v/v) trifluoroaceti cacid
- Purity (GC):Min. 96.0 %
- IR spectrum:complying
- Refractive index (n 20/D):1.385 - 1.389
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.02 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance 280 nm:Max. 0.01
- Absobance at 254 nm:Max. 0.01
- Absorbance at 225 nm:Max. 0.10
- Absorbance at 210 nm:Max. 0.40
- Absorbance 197 nm:Max. 1.0
- APHA:Max. 10
- Refractive Index:n20/D 1.387 (lit.)
- Autoignition Temperature:433 °F
Specification Test Results
About this item
:
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.9982 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:76-05-1
MDL Number:MFCD00011332