Order Entry
United States
ContactUsLinkComponent
Acetonitrile : Water (65:35)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile : Water (65:35)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ581-4
Supplier:  Avantor
CAS Number:  
Acetonitrile : Water (65:35)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ581-4
Supplier:  Avantor
CAS Number:  

About this item

Boiling Pt:67.7 °C
Density:0.8681 g/cm3
Flash Pt:8.3 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :GHS08
 :GHS02
 :GHS07
CAS Number:75-05-8
 :7732-18-5