

Dichloromethane : Methanol mixture (90:10)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ492-4
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ492-4
Specifications
About this item
:
Boiling Pt:40.56 °C
Density:1.2711 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS08
:GHS07