Acetonitrile : Water (70:30)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-05-8
:7732-18-5
Boiling Pt:67.7 °C
Density:0.8561 g/cm³ (25 °C)
Flash Pt:8.3 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
Frequently Bought Together