Order Entry
United States
ContactUsLinkComponent
Acetone ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Irritant
Flammable
Acetone ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Catalog # BJ414689-4X4L
CAS Number:  
undefined
Irritant
Flammable
Acetone ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Catalog # BJ414689-4X4L
Supplier Number:  414689-4X4L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    BJ414689-4X4L

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.5 %
  • Suitability for residue analysis:
    Max. 2 ng/l
  • Organic impurities as hexadecane (GC-FID):
    Max. 1 µg/l
  • Vapor Pressure:
    184 mmHg ( 20 °C)

Specification Test Results

About this item

 :
 :58.08 g/mol
Boiling Pt:56 °C
Melting Pt:–94.8 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :GHS07
 :GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765