Order Entry
United States
ContactUsLinkComponent
2-Propanol VLSI, Semiconductor grade (Honeywell 17604)
Flammable
Irritant
2-Propanol VLSI, Semiconductor grade (Honeywell 17604)
Certificates
undefined
Flammable
Irritant
2-Propanol VLSI, Semiconductor grade (Honeywell 17604)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Particles > 0,5 µm:
    Max. 250 P/ml
  • Assay (GC):
    Min. 99.8 %
  • Boiling range:
    81 - 83 °C
  • Density (D 20/4):
    0.785 - 0.787
  • Refractive index (n 20/D):
    1.3770 - 1.3780
  • Non-volatile matter:
    Max. 5 ppm
  • Water (Karl Fischer):
    Max. 500 ppm
  • Free acid (as C₂H₅COOH):
    Max. 10 ppm
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.02 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.05 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.05 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.02 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.02 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.1 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.02 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.02 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 1 ppm
  • Acetone (GC):
    Max. 100 ppm

Specification Test Results

Related Information

Frequently Bought Together