Acetonitrile : Water (95:5)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ403-4
Supplier: Avantor
CAS Number:
About this item
Boiling Pt:70 °C
Density:0.7892 g/cm3
Flash Pt:6.1 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:75-05-8
:7732-18-5