

Ammonium fluoride solution 40% VLSI, Semiconductor grade (Honeywell 17600)
Catalog # BJ40292H-5L
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:5 L
- Cat. No.:BJ40292H-5L
Specifications
Specification Test Results
- Particles > 0,5 µm:Max. 250 P/ml
- Assay:40 - 41 %
- Residue on ignition (SO₄):Max. 10 ppm
- pH (1 %,20°C):6.2 - 7.0
- Silver (Ag):Max. 0.05 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic, antimony (as As):Max. 0.03 ppm
- Gold (Au):Max. 0.02 ppm
- Boron (B):Max. 0.05 ppm
- Barium (Ba):Max. 0.01 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.02 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Iron (Fe):Max. 0.05 ppm
- Gallium (Ga):Max. 0.01 ppm
- Germanium (Ge):Max. 0.05 ppm
- Indium (In):Max. 0.01 ppm
- Potassium (K):Max. 0.05 ppm
- Lithium (Li):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 0.1 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Platinum (Pt):Max. 0.05 ppm
- Silicon (Si):Max. 200 ppm
- Tin (Sn):Max. 0.02 ppm
- Strontium (Sr):Max. 0.01 ppm
- Titanium (Ti):Max. 0.01 ppm
- Thallium (Tl):Max. 0.02 ppm
- Vanadium (V):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.01 ppm
- Ammonium hydrogen difluoride (NH₄HF₂):Max. 200 ppm
- Chloride (Cl):Max. 4 ppm
- Phosphate (PO₄):Max. 0.2 ppm
- Sulfate (SO₄):Max. 2 ppm
Specification Test Results
About this item
:
:37.04 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.110 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3287
ADR:6.1,III
:GHS05
:GHS06
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423