Order Entry
United States
ContactUsLinkComponent
N,N-Dimethylacetamide ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Irritant
Health
N,N-Dimethylacetamide ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # BJ38840-1L
CAS Number:  
undefined
Irritant
Health
N,N-Dimethylacetamide ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # BJ38840-1L
Supplier Number:  38840-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ38840-1L

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.436 - 1.439
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Non-volatile matter:
    Max. 0.005 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.008 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm

Specification Test Results

About this item

 :
 :87.12 g/mol
Boiling Pt:164-166 °C (1.013 hPa)
Boiling Pt:164…166 °C (1.013 hPa)
Melting Pt:-20 °C
Melting Pt:–20 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:11.5 %(V) (99 °C)
EINECS:204-826-4
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686