


Acetonitrile with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Catalog # BJ34678-4X4L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ34678-4X4L
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Acetic acid (CH₃COOH):0.093 - 0.107 %
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 210 nm:Min. 20 %
- Transmittance at 230 nm:Min. 50 %
- Transmittance at 254 nm:Min. 90 %
- Curve course (UV):complying
- HPLC Gradient at 254 nm:Max. 50 mAU
- LC-MS application:complying
Specification Test Results
About this item
:
Boiling Pt:83 °C (main component)
Melting Pt:–46 °C (main component)
Density:0.7820 g/cm³ (20 °C)
Flash Pt:19 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:75-05-8
MDL Number:MFCD00001878