Acetonitrile with 0.1% formic acid and 0.01% trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
BJ34676-4X4L
:
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ34676-4X4L
- assay (GC):Min. 99.5 %
- assay of formic acid:0.093 - 0.107 Vol.%
- assay trifluoroacetic acid:0.009 - 0.011 Vol.%
- calcium (Ca):Max. 0.5 ppm
- potassium (K):Max. 0.5 ppm
- magnesium (Mg):Max. 0.5 ppm
- sodium (Na):Max. 2 ppm
- transmittance at 210 nm:Min. 5 %
- transmittance at 230 nm:Min. 15 %
- transmittance from 254 nm:Min. 90 %
- HPLC-gradient at 254 nm:Max. 50 mAU
- LC-MS application:complying
:
:82 °C (1.013 hPa) (main component)
:–46 °C (main component)
:0.777 g/cm³ (25 °C)
:6.1 °C
:Ambient
:1648
:3,II
:GHS08
:GHS07
:GHS02
:64-18-6