Order Entry
United States
ContactUsLinkComponent
 
Water with 0,1% ammonium acetate, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Catalog # BJ34674-4X4L
CAS Number:  
Water with 0,1% ammonium acetate, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Catalog # BJ34674-4X4L
Supplier Number:  34674-4X4L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    BJ34674-4X4L

Specifications

Specification Test Results

  • Assay of ammonium acetate:
    0.093 - 0.107 %
  • pH (20 °C):
    6.8 - 7.2
  • Calcium (Ca):
    Max. 0.5 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Magnesium (Mg):
    Max. 0.5 ppm
  • Sodium (Na):
    Max. 2 ppm
  • Transmittance at 210 nm:
    Min. 20 %
  • Transmittance at 230 nm:
    Min. 90 %
  • Transmittance from 254 nm:
    Min. 99 %
  • HPLC Gradient at 210 nm:
    Max. 50 mAU
  • HPLC Gradient at 254 nm:
    Max. 10 mAU
  • LC-MS application:
    complying

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Density:0.9971 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:631-61-8