Order Entry
United States
ContactUsLinkComponent
Acetonitrile : Water (30:70)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile : Water (30:70)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ2008-4
Supplier:  Avantor
CAS Number:  
Acetonitrile : Water (30:70)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJ2008-4
Supplier:  Avantor
CAS Number:  

About this item

Boiling Pt:70 °C
Density:0.9509 g/cm3
Flash Pt:13.3 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :GHS08
 :GHS07
 :GHS02
CAS Number:75-05-8
 :7732-18-5