ContactUsLinkComponent
Honeywell™ Circular Chart Recorder, Industrial Container Services
Honeywell™ Circular Chart Recorder, Industrial Container Services
Catalog # MSPP-DR45AT111
Irregular Voltage
Honeywell™ Circular Chart Recorder, Industrial Container Services
Catalog # MSPP-DR45AT111
Supplier Number:  26949
Irregular Voltage
Specifications
  • Description:
    12" Circular chart recorder, 4 Analog inputs
  • Cat. No.:
    MSPP-DR45AT111
Specifications
About this item
The Circular chart recorder is a microprocessor-based recorder suitable to meet a wide range of industry requirements. It comes with a digital display and easy-to-use operator interface, and is available in many models to cater to the specific needs of different applications such as flow and HTST dairy processes.
Circular chart recorder aimed at applications:
  • Food processing
  • Pharmaceuticals
  • Heat treatment
  • Environmental testing
  • Manufactured goods and water/waste water treatment
It creates its own chart, thereby eliminating the need for an inventory of preprinted charts and providing flexibility to the user.