ContactUsLinkComponent
igScript™ One Step RT-PCR Kits, Intact Genomics
igScript™ One Step RT-PCR Kits, Intact Genomics
Catalog # 103753-214
Supplier:  Intact Genomics
CAS Number:  
undefined
igScript™ One Step RT-PCR Kits, Intact Genomics
Catalog # 103753-214
Supplier:  Intact Genomics
Supplier Number:  4211
CAS Number:  
Specifications
  • Description:
    igScript™ One Step RT-PCR Kit
  • Size:
    100 Reactions
  • Cat. No.:
    103753-214
Specifications
About this item
igScript™ one step RT-PCR kit combines two powerful mixtures: igScript™ reverse transcriptase and hot start Taq 2X master mix providing improved PCR efficiency, wider dynamic range, superior sensitivity and specificity. The two mixtures require minimal handling during reaction setup and offer consistent and robust RT-PCR reactions.
  • Robust and active for cDNA synthesis at temperatures up to 55 °C
  • Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA
  • Useful for cDNA synthesis followed by gene expression data validation
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Hot start Taq 2X master mix
igScript™ reverse transcriptase is a recombinant MMLV reverse transcriptase with reduced RNase H activity and increased thermostability. The kit is highly efficient at producing full-length cDNA from long RNA templates at temperatures between 42 to 55 °C.
Hot start Taq 2X master mix is a ready-to-use cocktail containing all components except primers and template, for the amplification and detection of DNA in PCR. The hot start Taq 2X master mix contains standard buffer, chemically-modified hot start Taq polymerase, MgCl₂, dNTPs and stabilizers. The amplification step features a high quality hot start Taq DNA Polymerase which offers higher fidelity and better amplification.
Delivery: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.