i7™ Hhot start high-fidelity DNA polymerase 2X master mix is ready to use premix which contains hot start high-fidelity DNA polymerase, dNTPs,MgCl₂, PCR enhancers and stabilizers with optimized proprietary reaction buffer. i7™ hot start high-fidelity DNA polymerase 2X master mix has the high-fidelity, sensitivity and processivity with an error rate ~2.8×102 fold lower than Taq DNA polymerase, and significantly lower than the error rates of other proofreading enzymes.
- Useful for long and difficult DNA amplification, high fidelity PCR, and cloning
- Supplied as convenient master mix
- Efficient for amplifying high GC template DNA with magic enhancer
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 5X magic enhancer
Hot start i7™ high-fidelity DNA polymerase 2X master mix is ready to use premix which contains hot start i7™ high-fidelity DNA polymerase, dNTPs, MgCl₂, PCR enhancers and stabilizers with optimized proprietary reaction buffer. Hot start i7™ High-fidelity DNA polymerase is a chemically engineered, heat stable DNA polymerase which has 5' - 3' polymerase and 3' - 5' exonuclease (proof reading) activities. This enzyme has high-fidelity, sensitivity and processivity with an error rate ~2.8×10^2-fold lower than Taq DNA polymerase, and significantly lower than other proo freading enzymes in the marketplace.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Environmentally Preferable: