igTaq DNA Polymerase 2X Master Mix, Intact Genomics
103753-148
:
- Description:igTaq DNA Polymerase 2X Premix
- Size:500 Reactions (12.5 ml)
- Cat. No.:103753-148
High quality Taq DNA polymerase 2X master mix useful for routine PCR applications.
- Useful for routine PCR, primer extension, colony PCR and more
- Formulated for amplifying long target DNA
- Supplied as convenient master mix
- Efficient for amplifying high GC template DNA with magic enhancer
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- Taq DNA polymerase 2X premix with dye
- 5X magic enhancer
Intact Genomics Taq DNA Polymerase 2X premix with dye is ready to use, containing Taq DNA polymerase, dNTPs, MgCl₂, and stabilizers. Taq DNA polymerase is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity. This product is supplied with 10X PCR reaction buffer, containing MgCl₂, which produces a final Mg²⁺ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3' ends.
: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.