Order Entry
United States
ContactUsLinkComponent
 
Test Tube pH Pen, Flat, Sper Scientific
  76333-862
undefined
Test Tube pH Pen, Flat, Sper Scientific
  76333-862
 :  850066

 

  • Description:
    Test Tube pH Pen with Flat Tip
  • Compliance:
    IP65
  • Dimensions:
    9×13/8×1"
  • Sensor type:
    Flat pH sensor 5” × ½" (125 × 12 mm)
  • Temperature Compensation:
    Automatic
  • Weight:
    2.3 oz.
  • pH Accuracy:
    ±0.1
  • pH Calibration:
    3 points
  • pH Range:
    2 to 12
  • pH Resolution:
    0.01
  • Cat. No.:
    76333-862

 

 

Fits standard test tubes 16×50 mm or larger.

  • Flat tip probe for meat, leather, paper, skin, and other surface pH measurements
  • Highly stable and accurate readings with a large LCD display
  • Designed for simple one-hand operation
  • Features hold function, and auto power off with low battery indicator

Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize some other non-standard buffers as well.

The case is IP 65 waterproof and will float if accidentally dropped into the water.

 : Comes with two button-cell batteries and a built-in electrode.