T4 UvsX DNA Recombinase
Supplier: INTACT GENOMICS, INC MS
Certificates
About this item
Offers the largest collection of T4 Enzymes and proteins in the industry.
- Extremely high purity
- Variety of uses including RPA optimization
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 10X UvsX recombinase reaction buffer
Homologous recombination is important for the error-free repair of DNA double-strand breaks and for replication fork restart. Recombinases of the RecA/RAD51 family perform the central catalytic role in this process. UvsX recombinase is the RecA/Rad51 ortholog of bacteriophage T4. T4 UvsX DNA recombinase and other recombinases form presynaptic filaments on ssDNA that are activated to search for homology in dsDNA and to perform DNA strand exchange.
Delivery: All our products are shipped on dry ice.
Specifications
- Enzyme Name:T4 UvsX DNA Recombinase
- Concentration:5 μg/μl