About this item
igScript™ reverse transcriptase, is a recombinant, Moloney Murine Leukemia virus (MMLV) reverse transcriptase with reduced RNase H activity.
- Recombinant MMLV reverse transcriptase with greatly reduced RNase H activity
- Active at temperatures up to 55 °C
- Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA
- Useful for first strand cDNA synthesis for PCR or RT-PCR and gene expression data validation by using RT-PCR
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 10X igScript™ RT reaction buffer
igScript™ reverse transcriptase (igScript RT) is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates. igScript RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR analysis and other applications.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:Moloney murine leukemia virus
- Enzyme Name:igScript™ Reverse Transcriptase
- Concentration:200 U/µl