Cas9 Nuclease (from Streptococcus pyogenes)
Cas9 nuclease is the purified recombinant Streptococcus pyogenes Cas9 enzyme containing a nuclear localization signal (NLS) at the C-terminal for targeting to the nucleus. This enzyme is designed to perform CRISPR/Cas9-mediated genome editing.
- Ideal for in vitro digestion of dsDNA
- Extremely high purity
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- Cas9 nuclease
- 10X Cas9 nuclease reaction buffer
The physical purity of this enzyme is ≥98% as assessed by SDS-PAGE with Coomassie® blue staining.
: All our products are shipped with dry ice.
- Source:Streptococcus pyogenes
- Enzyme Name:Cas9 Nuclease
- Concentration:1600 ng/µl
Frequently Bought Together