Order Entry
United States
ContactUsLinkComponent
Visual Kit, Nitrate, CHEMets®
Visual Kit, Nitrate, CHEMets®
Catalog # 10736-448
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Visual Kit, Nitrate, CHEMets®
Catalog # 10736-448
Supplier:  CHEMetrics
Supplier Number:  K-6905
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. No.:
    10736-448
  • Range:
    0 - 3.4 ppm as N
  • No. of tests:
    30

Specifications

About this item

CHEMetrics easy-to-use snap-and-read water analysis test kit.

  • Analysis time: 12 minutes
  • Zinc reduction method

Comparator gradations: 0, 0.3, 0.6, 0.9, 1.3, 1.7, 2.2, 2.8, 3.4 ppm

Delivery: Nitrate Test Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, Acidifier Solution, reaction tube and cap, 25 mL sample cup, and instructions.
Caution: The refill and color comparator contained in this kit have shelf-lives of 12 months.