Specifications
- Description:MIDORI Green Xtra
- Size:1 ml
- Environmentally Preferable:
- Cat. No.:103778-502
Specifications
About this item
Midori Green Xtra — Safe DNA Stain Non-carcinogenic, Non-toxic, and Über-sensitive
- Put it in your sample not your gel
MIDORI Green Xtra is the newest addition to the family of MIDORI Green DNA stains from Nippon Genetics. Xtra is a highly sensitive green fluorescent stain for a safe visualization of DNA and RNA in agarose gels using either blue LEDs or our superior Blue/Green LED Gel Illuminators and Imagers. It is markedly superior to the traditional nucleic acid stain ethidium bromide (EtBr) in just about every way. Agarose gels stained with Xtra have an ultra-low background fluorescence, which makes the identification of even tiny amounts of DNA very easy. Free samples are available, so see for yourself how MIDORI Green Xtra provides safe detection with unbeatable sensitivity.