Chemically competent DL39 (DE3) E. coli cells are specific for transformation and protein expression in order to uniformly and specifically label e.g. phenylalanine or leucine residues. DL39 (DE3) can also be used to reduce NMR cross-labeling via. transaminase activity for valine, leucine, isoleucine, aspartate, phenylalanine, tyrosine and tryptophan residues.
- DL39 (DE3) chemically competent cells provide the highest transformation efficiency in the industry
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- Includes DL39 (DE3) competent cells, pUC19 control DNA and recovery medium
Genomic features deficient in the aromatic (TyrB), branched-chain (JIvE), and aspartate (AspC) transaminases. Modified to contain the T7 expression system.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 103753-100
- 103753-098
- Genotypes:
- F-, λ-, aspC13, fnr-25, rph-1, ilvE12, tyrB507,λDE3
- F-, λ-, aspC13, fnr-25, rph-1, ilvE12, tyrB507,λDE3