ContactUsLinkComponent
BL21 Chemically Competent Cells, Intact Genomics
BL21 Chemically Competent Cells, Intact Genomics
Catalog # 103753-088
Supplier:  Intact Genomics
undefined
BL21 Chemically Competent Cells, Intact Genomics
Catalog # 103753-088
Supplier:  Intact Genomics
Supplier Number:  1041-24
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Volume:
    24× 50 µl
  • Description:
    BL21 Chemically Competent Cells
  • Genotypes:
    F- dcm ompT hsdS(rB- mB-) gal [malB+]K-12(λS)
  • Cat. No.:
    103753-088
Specifications
About this item
BL21 chemically competent cells are suitable for transformation and routine protein expression from non-T7 vectors.
  • Intact genomics (IG) BL21 chemically competent cells provide the highest transformation efficiency (≥1×108 Cfu/µg pUC19 DNA) in the industry
  • Available as single-use vials
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • Includes BL21 competent cells, pUC19 control DNA and recovery medium
Genomic features a widely used host background, routine non-T7 vector expression, deficient in both lon and ompT proteases and resistance to phage T1 (fhuA2).
Delivery: All our products are shipped with dry ice.