durAstatic® Dissipative HDPE Bottles, with Black Screw Cap, Menda
: Desco
HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry.
- No migratory additives
- Embossed with ESD protective symbol
- Vessel of 6 oz. includes measurements on one side to monitor fluid level
- Black screw cap is insulative
Available in four colors for easy identification in the workplace, HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry. No migratory additives are used, reducing the chance for contamination from the bottle.
Black screw cap easily allows opening and reclosing to prevent contents from spilling. The cap is insulative.
: Long lasting dissipative properties: Rtt: 10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.
Frequently Bought Together