Wright-Giemsa stain solution
Supplier: Azer Scientific Inc.
Jump to:
About this item
Wright-Giemsa stain is a modified Romanowsky stain used for staining the cellular elements in blood and bone marrow
- Provides excellent differential staining results
- Stains RBC inclusion bodies, parasites and other organisms in blood smears
- Sutiable for automated or manual staining methods
Synonyms: Wright-Giemsa Solution
- MDL Number:MFCD00132593
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Wright-Giemsa stain is a modified Romanowsky stain used for staining the cellular elements in blood and bone marrow
- Provides excellent differential staining results
- Stains RBC inclusion bodies, parasites and other organisms in blood smears
- Sutiable for automated or manual staining methods
Synonyms: Wright-Giemsa Solution
- MDL Number:MFCD00132593
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot Numbersds_es921
SDS
VWR GHS Compliance Letter (US)



