10 L Unjacketed Filter Reactor Systems, Air Motor, Chemglass
89209-346
Small Business Enterprise
- Capacity:10 L
- Description:UnJacketed Cylindrical Process Filter Reactor System Complete
- Motor type:Air motor
- Cat. No.:89209-346
- Supplier no.:CG-1958-F811
Unjacketed filter glass reactors feature autocenter stand.
- Complete reactor system on open air, autocenter stand
- 150 °C maximum temperature limit for filter base
- Filter base eliminates material transfers and significantly reduces losses
Jacketed filter glass reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.