Coil Condenser Inserts, Type C, Chemglass
Catalog # 89028-426
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Coil Condenser Insert, Type C
- Cat. No.:89028-426
- Supplier no.:CG-1332-10
Specifications
About this item
Condensers are for use with either tap water or a chilled circulator.
- Insert allows the conversion of a C type dry ice condenser
- For use with either tap water or a chilled circulator
Inlet and outlet are GL-14 threads and supplied complete with caps and detachable hose connections. Also supplied with a PTFE centering ring.