ContactUsLinkComponent
Support Stands, Stainless Steel, Chemglass
Support Stands, Stainless Steel, Chemglass
Catalog # 33501-428
Supplier:  Chemglass
Small Business Enterprise
undefined
Support Stands, Stainless Steel, Chemglass
Catalog # 33501-428
Supplier:  Chemglass
Supplier Number:  CG-1539-S-100
Small Business Enterprise
Specifications
  • Description:
    Stainless Steel Support, Floor Model
  • Environmentally Preferable:
  • Cat. No.:
    33501-428
  • Supplier no.:
    CG-1539-S-100
Specifications
About this item
Floor style stainless steel support stands.
  • Made to accept 12 and 22 liter CG-1539 half jacketed flasks
  • Top of stand has a 39" reinforced riser to support overhead stirrers, clamps and so on
Stand is 67" overall height with a depth of 22" ring has a diameter of 81/2 " I.D. and is 28" above floor level.