Adapter, Rotary Evaporator Bump Trap, R200 and R205, Chemglass
Catalog # 33501-708
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Vapor Tube/Trap Adapter, 24/40
- Cat. No.:33501-708
- Supplier no.:CG-1317-B-10
Specifications
About this item
Vapor tube/bump trap adapter is designed to be used with the Buchi® R-200/205 rotary evaporators.
- This adapter combines a vapor tube adapter and a bump trap
- Tapered sidewalls permit continuous washing with condensed solvent
- While preventing vapor cooling points along the path of the condenser
- The upper stem has two large holes flush with the lower end
- Allows solvent vapor to flow freely
- This portion of the trap is easy to clean
Adapters have a 50 mm outer Ø top flange with a tapered interior to fit the Büchi® sealing ring. Lower portion of the adapter has the listed standard taper inner joint.
Note: Items will replace the Buchi® part numbers as listed. Chemglass items are to be used with the Keck clip, not the Buchi Combi-clip.
Delivery: Each adapter is supplied complete with the corresponding size keck clip.