Adapters, Rotary Evaporator Bump Trap, Fritted, R200 and R205, Chemglass
Catalog # 33501-712
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Vapor Tube/Trap Adapter, Fritted
- Environmentally Preferable:
- Cat. No.:33501-712
- Joint size:24/40
- Supplier no.:CG-1317-C-10
Specifications
About this item
Vapor tube/bump trap adapter is designed to be used with the Büchi® R-200/205 rotary evaporators.
- This adapter combines a vapor tube adapter and a bump trap
- Trap has an extra coarse fritted disc sealed between the lower inner joint and the top flange
- Adapters have a 50 mm OD top flange with a tapered interior to fit the Büchi® sealing ring
Lower portion of the adapter has the listed standard taper inner joint. Each adapter is supplied complete with the corresponding size Keck clip.