Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant I-TAC (CXCL11) (from E. coli)
 :  Peprotech
 
Human Recombinant I-TAC (CXCL11) (from E. coli)
 :  Peprotech

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological Activity:
    Determined by its ability to chemoattract human T-cells activated with IL-2.
  • Gene ID:
    6373
  • Animal-Free:
    Y
  • Protein Synonyms:
    Interferon Inducible T-Cell alpha Chemokine|B-R1
  • Protein/Peptide Name:
    I-TAC (CXCL11)
  • Purity:
    98%
  • Molecular Weight:
    8.3 kDa
  • Sequence:
    FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together