Orders LinkContactUsLinkComponent
Desktop Aquaponics System
Desktop Aquaponics System
Supplier:   R&D AQUAFARMS SE
Desktop Aquaponics System
Supplier:  R&D AQUAFARMS SE
Certificates
Desktop Aquaponics System
Desktop Aquaponics System
2 Options Available Below
Product Details & Documents
A compact system to easily display principles.
  • Small footprint
  • Includes tank
  • Easily accessed for cleaning
  • Great size for setup on a cart
Easily show the principles of aquaponics in this compact system. The system uses a space-saving five gallon aquarium (included) utilizing a simple airlift pump. The water flows through the filter media along to the plants. The filtered water then returns to the fish. This sized system can be used to grow a variety of herbs and lettuces. The LED grow light (optional) is positioned directly over the grow raft. The height can be adjusted as the plants grow. Add water along with choice of fish and seeds. Then watch the ecosystem grow.
The top can be removed for easy cleaning. A small heater can be added for warm water fish.
Product Family Options
Product Information
  • Availability
    Price
  • 470320-464
    Desktop Aquaponics
    Desktop Aquaponics
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $416.95
    $416.95
    Kit of 1
       $416.95
    Minimum order quantity is 1
    Supplier Number: KCTA
  • 470320-466
    Desktop Aquaponics with Light
    Desktop Aquaponics with Light
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $701.00
    $701.00
    Kit of 1
       $701.00
    Minimum order quantity is 1
    Supplier Number: KCTA-L