Order Entry
United States
Orders LinkContactUsLinkComponent
Solar Panel Demonstration, Eisco Scientific

Solar Panel Demonstration, Eisco Scientific

Solar Panel Demonstration
Catalog # 470230-442
Supplier:   Wards
Supplier #:  PH1320A
Solar Panel Demonstration, Eisco Scientific
Catalog # 470230-442
Supplier:  Wards
Solar Panel Demonstration, Eisco Scientific
Solar Panel Demonstration, Eisco Scientific

Solar Panel Demonstration

Price
$60.35
Log in for availability
Each
Product Details & Documents

Compact demonstration of alternative energy.

  • Six solar cells, connected in series on clear base
  • 12 V output, 130 mA current
  • Plugs of 4 mm

Designed for use in experiments related to the transfer of energy and is an excellent teaching tool to introduce students to the fundamentals of one of the most critical energy systems of our future.

The unit itself measures 8.5×6×1.5" and each solar cell measures 3×1". Designed with two 1.4" jacks.

Ordering information: Requires 4 mm leads, not included.

Specifications
  • Description:
    Solar Panel Demonstration