ContactUsLinkComponent
Homeowner's Retractable Utility Knife, Stanley
Homeowner's Retractable Utility Knife, Stanley
Catalog # 76252-532
Supplier:  TESTEQUITY LLC TE
Homeowner's Retractable Utility Knife, Stanley
Catalog # 76252-532
Supplier:  TESTEQUITY LLC TE
Supplier Number:  10-175
Specifications
  • Color:
    Gray
  • Description:
    Retractable Blade Utility Knife
  • Cat. No.:
    76252-532
  • Blade length:
    61.9 mm (2⁷⁄₁₆")
  • Handle length:
    61/8"
Specifications
About this item
The homeowner's retractable utility knife is lightweight and economical.
  • Three-position retractable blade
  • Interlocking overlap nose secures blade safely between knife halves
Indispensable tool for a do-it-yourselfer's tool box, shop cabinet, workbench, tool belt or kitchen junk drawer.