ContactUsLinkComponent
Cascade® Automatic Dishwasher Powder
Cascade® Automatic Dishwasher Powder
Catalog # 10099-524
undefined
Cascade® Automatic Dishwasher Powder
Catalog # 10099-524
Supplier Number:  PGC59535CT
Specifications
  • Cat. No.:
    10099-524
  • Packaging:
    Box
  • Form:
    Powder
  • Supplier no.:
    PGC59535
Specifications
About this item
For under-counter dishwashers and cabinet-type automatic utensil washers.
  • For use on ceramics, enamel, fiberglass, glass, metal, plastic
  • Gluten-free
  • Cleans cooked-on grease, food particles, oils, organic matter, soil
Leaves dishes virtually spotless.
Certifications: Kosher Certified.