Order Entry
United States
ContactUsLinkComponent
 
PURELL® Hand Sanitizing Wipes
  10714-058
undefined
PURELL® Hand Sanitizing Wipes
  10714-058
 :  GOJ911802

 

  • Color:
    White
  • Length:
    8"
  • Cat. No.:
    10714-058
  • Packaging:
    Refill Pouch
  • Supplier no.:
    GOJ9118-02
  • No. of towels/wipes per container:
    1200

 

 

Kills more than 99.99% of most common germs that may cause illness.

  • General purpose/hand washing
  • Removes light soils and dirt from hands
  • Moisturizers keep skin soft
  • Unscented
  • For use with 9115-DS1C

Ideal for offices, restaurants, health clubs or anywhere germs may be.