Total Dissolved Solids Meter
Catalog # 470315-074
Supplier: Avantor
Specifications
- Description:TDS Meter
- Cat. No.:470315-074
Specifications
About this item
The P773 is a wide-range, pocket-sized total dissolved solids (TDS) meter perfect for use in field environments.
Having a rugged, waterproof housing, and replaceable electrodes, this meter will give long durable service. With 10 ppm resolution and ±2% FS accuracy, you will have high quality results at your fingertips. Automatic temperature compensation (ATC) adjusts for changes in TDS due to temperature.